Assay Details
| Gene Symbol: Nfatc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010901.2 | NP_035031.2 | 5 - 6 | 2070 | 127 | |
| GenBank mRNA | AK131919.1 | - | 3 - 4 | 482 | 127 | |
| AK134054.1 | - | 5 - 6 | 1989 | 127 | ||
| AK147708.1 | - | 5 - 6 | 2040 | 127 | ||
| AK220187.1 | - | 5 - 6 | 1842 | 127 | ||
| BC021835.1 | - | 2 - 3 | 274 | 127 | ||
| BC120900.1 | - | 5 - 6 | 1789 | 40085687 | 127 | |
| BC120901.1 | - | 5 - 6 | 1789 | 40085688 | 127 | |
| D85612.1 | - | 5 - 6 | 1786 | 127 | ||
| EU887626.1 | - | 5 - 6 | 1758 | 127 | ||
| EU887627.1 | - | 5 - 6 | 1758 | 127 | ||
| EU887629.1 | - | 5 - 6 | 1782 | 127 | ||
| EU887630.1 | - | 5 - 6 | 1782 | 127 | ||
| U28807.1 | - | 5 - 6 | 1749 | 127 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nfatc3
Genomic Map