Assay Details
| Gene Symbol: Nfatc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010901.2 | NP_035031.2 | 7 - 8 | 2257 | 86 | |
| GenBank mRNA | AK131919.1 | - | 5 - 6 | 669 | 86 | |
| AK134054.1 | - | 7 - 8 | 2176 | 86 | ||
| AK147708.1 | - | 7 - 8 | 2227 | 86 | ||
| AK220187.1 | - | 7 - 8 | 2029 | 86 | ||
| BC021835.1 | - | 4 - 5 | 461 | 86 | ||
| BC120900.1 | - | 7 - 8 | 1976 | 40085687 | 86 | |
| BC120901.1 | - | 7 - 8 | 1976 | 40085688 | 86 | |
| EU887626.1 | - | 7 - 8 | 1945 | 86 | ||
| EU887627.1 | - | 7 - 8 | 1945 | 86 | ||
| EU887629.1 | - | 7 - 8 | 1969 | 86 | ||
| EU887630.1 | - | 7 - 8 | 1969 | 86 | ||
| U28807.1 | - | 7 - 8 | 1936 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nfatc3
Genomic Map