Assay Details
| Gene Symbol: Nfatc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010901.2 | NP_035031.2 | 8 - 9 | 2394 | 79 | |
| GenBank mRNA | AK134054.1 | - | 8 - 9 | 2313 | 79 | |
| AK147708.1 | - | 8 - 9 | 2364 | 79 | ||
| AK220187.1 | - | 8 - 9 | 2167 | 79 | ||
| BC021835.1 | - | 5 - 6 | 598 | 79 | ||
| BC120900.1 | - | 8 - 9 | 2113 | 40085687 | 79 | |
| BC120901.1 | - | 8 - 9 | 2113 | 40085688 | 79 | |
| D85612.1 | - | 8 - 9 | 2110 | 79 | ||
| EU887626.1 | - | 8 - 9 | 2082 | 79 | ||
| EU887627.1 | - | 8 - 9 | 2082 | 79 | ||
| EU887629.1 | - | 8 - 9 | 2106 | 79 | ||
| EU887630.1 | - | 8 - 9 | 2106 | 79 | ||
| U28807.1 | - | 8 - 9 | 2073 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nfatc3
Genomic Map