Assay Details
| Gene Symbol: Nfatc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010901.2 | NP_035031.2 | 1 - 2 | 396 | 158 | |
| GenBank mRNA | AK039929.1 | - | 1 - 2 | 313 | 158 | |
| AK051602.1 | - | 1 - 2 | 354 | 158 | ||
| AK134054.1 | - | 1 - 2 | 315 | 158 | ||
| AK147708.1 | - | 1 - 2 | 367 | 158 | ||
| AK155087.1 | - | 1 - 2 | 366 | 158 | ||
| AK220187.1 | - | 1 - 2 | 168 | 158 | ||
| BC120900.1 | - | 1 - 2 | 115 | 40085687 | 158 | |
| BC120901.1 | - | 1 - 2 | 115 | 40085688 | 158 | |
| EU887629.1 | - | 1 - 2 | 108 | 158 | ||
| EU887630.1 | - | 1 - 2 | 108 | 158 | ||
| EU887631.1 | - | 1 - 2 | 108 | 158 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nfatc3
Genomic Map