Assay Details
| Gene Symbol: Cdh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037809.5 | NP_001032898.1 | 11 - 12 | 1656 | 120 | |
| NM_007665.3 | NP_031691.1 | 11 - 12 | 1653 | 120 | ||
| GenBank mRNA | AK031265.1 | - | 11 - 12 | 1596 | 120 | |
| AK045041.1 | - | 11 - 12 | 1616 | 120 | ||
| AK164577.1 | - | 11 - 12 | 1626 | 120 | ||
| BC052189.1 | - | 9 - 10 | 1344 | 120 | ||
| BC098459.1 | - | 11 - 12 | 1562 | 30079668 | 120 | |
| X06340.1 | - | 11 - 12 | 1594 | 120 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdh3
Genomic Map