Assay Details
| Gene Symbol: Afmid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027827.3 | NP_082103.1 | 6 - 7 | 493 | 67 | |
| GenBank mRNA | AF399717.1 | - | 6 - 7 | 488 | 67 | |
| AK018563.1 | - | 4 - 5 | 429 | 67 | ||
| AY099479.1 | - | 6 - 7 | 488 | 67 | ||
| BC024452.1 | - | 6 - 7 | 464 | 67 | ||
| BC043309.1 | - | 6 - 7 | 459 | 67 | ||
| BC052507.1 | - | 2 - 3 | 117 | 67 | ||
| BC066780.1 | - | 6 - 7 | 472 | 30301832 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Afmid
Genomic Map