Assay Details
| Gene Symbol: Afmid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027827.3 | NP_082103.1 | 8 - 9 | 673 | 69 | |
| GenBank mRNA | AF399717.1 | - | 8 - 9 | 668 | 69 | |
| AK018563.1 | - | 6 - 7 | 609 | 69 | ||
| AY099479.1 | - | 8 - 9 | 668 | 69 | ||
| BC024452.1 | - | 8 - 9 | 644 | 69 | ||
| BC043309.1 | - | 8 - 9 | 639 | 69 | ||
| BC066780.1 | - | 8 - 9 | 652 | 30301832 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Afmid
Genomic Map