Assay Details
| Gene Symbol: Necab3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021546.3 | NP_067521.2 | 9 - 10 | 765 | 87 | |
| GenBank mRNA | AB039948.1 | - | 7 - 8 | 586 | 87 | |
| AK044296.1 | - | 9 - 10 | 757 | 87 | ||
| AK158225.1 | - | 9 - 10 | 767 | 87 | ||
| BC126874.1 | - | 7 - 8 | 579 | 40044848 | 87 | |
| BC126875.1 | - | 7 - 8 | 579 | 40044849 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Necab3
Genomic Map