Assay Details
| Gene Symbol: Necab3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021546.3 | NP_067521.2 | 1 - 2 | 114 | 230 | |
| GenBank mRNA | AK013520.1 | - | 1 - 2 | 113 | 230 | |
| AK044296.1 | - | 1 - 2 | 106 | 230 | ||
| AK158225.1 | - | 1 - 2 | 116 | 230 | ||
More Information
Set Membership: |
|

Necab3
Genomic Map