Assay Details
| Gene Symbol: Gm16208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK028089.1 | - | 2 - 3 | 180 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
syntrophin, basic 2 |
Sntb2 | Snt2 | -15.5kb | Anti-sense | Chr. 8 - 106935750 - 107014192 on NCBI Build GRCm38 |
| Downstream | vacuolar protein sorting 4a (yeast) | Vps4a |
4930589C15Rik, AI325971, AW553189 |
1.0kb | Anti-sense | Chr. 8 - 107031326 - 107045756 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm16208
Genomic Map