Assay Details
| Gene Symbol: Gm32643 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK050336.1 | - | 3 - 4 | 309 | 106 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 1903 | Mir1903 |
Mirn1903, mmu-mir-1903 |
-96.2kb | Anti-sense | Chr. 8 - 128359241 - 128359320 on NCBI Build GRCm38 |
| Downstream | integrin beta 1 (fibronectin receptor beta) | Itgb1 |
4633401G24Rik, AA409975, AA960159, CD29, ENSMUSG00000051907, Fnrb, Gm9863, gpIIa |
227.3kb | Anti-sense | Chr. 8 - 128685284 - 128733579 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm32643
Genomic Map