Assay Details
| Gene Symbol: Ncoa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008679.3 | NP_032705.2 | 16 - 17 | 3334 | 78 | |
| GenBank mRNA | AF000581.1 | - | 15 - 16 | 3211 | 78 | |
| AK142306.1 | - | 16 - 17 | 3239 | 78 | ||
| AK156345.1 | - | 16 - 17 | 3299 | 78 | ||
| BC086482.1 | - | 16 - 17 | 3304 | 30104508 | 78 | |
| BC093478.1 | - | 13 - 14 | 2965 | 78 | ||
| BC141177.1 | - | 15 - 16 | 3150 | 9055789 | 78 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ncoa3
Genomic Map