Assay Details
| Gene Symbol: Ncoa3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008679.3 | NP_032705.2 | 4 - 5 | 472 | 102 | |
| GenBank mRNA | AF000581.1 | - | 3 - 4 | 361 | 102 | |
| AK137507.1 | - | 4 - 5 | 458 | 102 | ||
| AK142306.1 | - | 4 - 5 | 377 | 102 | ||
| AK156345.1 | - | 4 - 5 | 450 | 102 | ||
| BC086482.1 | - | 4 - 5 | 442 | 30104508 | 102 | |
| BC093478.1 | - | 1 - 2 | 103 | 102 | ||
| BC141177.1 | - | 3 - 4 | 303 | 9055789 | 102 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ncoa3
Genomic Map