Assay Details
| Gene Symbol: Ptpn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011201.3 | NP_035331.3 | 1 - 2 | 228 | 85 | |
| GenBank mRNA | AK149979.1 | - | 1 - 2 | 184 | 85 | |
| AK150322.1 | - | 1 - 2 | 184 | 85 | ||
| AK153088.1 | - | 1 - 2 | 167 | 85 | ||
| AK156281.1 | - | 1 - 2 | 189 | 85 | ||
| AK157431.1 | - | 1 - 2 | 229 | 85 | ||
| AK165567.1 | - | 1 - 2 | 193 | 85 | ||
| AK166020.1 | - | 1 - 2 | 184 | 85 | ||
| AK168255.1 | - | 1 - 2 | 166 | 85 | ||
| AK168772.1 | - | 1 - 2 | 183 | 85 | ||
| AK170620.1 | - | 1 - 2 | 184 | 85 | ||
| AK171380.1 | - | 1 - 2 | 184 | 85 | ||
| BC010191.1 | - | 1 - 2 | 146 | 3156854 | 85 | |
| M97590.1 | - | 1 - 2 | 144 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn1
Genomic Map