Assay Details
| Gene Symbol: Ogfod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001093757.1 | NP_001087226.1 | 10 - 11 | 1379 | 68 | |
| NM_177767.4 | NP_808435.3 | 11 - 12 | 1508 | 68 | ||
| GenBank mRNA | AK029629.1 | - | 11 - 12 | 1510 | 68 | |
| AK122532.1 | - | 11 - 12 | 1412 | 68 | ||
| AK156765.1 | - | 11 - 12 | 1478 | 68 | ||
| AK164189.1 | - | 10 - 11 | 1266 | 68 | ||
| AK166343.1 | - | 11 - 12 | 1431 | 68 | ||
| AK202999.1 | - | 2 - 3 | 194 | 68 | ||
| BC040267.1 | - | 10 - 11 | 1339 | 5364674 | 68 | |
| EL608450.1 | - | 2 - 3 | 68 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Ogfod1
Genomic Map