Assay Details
| Gene Symbol: Gcnt7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039560.3 | NP_001034649.1 | 1 - 2 | 110 | 125 | |
| GenBank mRNA | AK039414.1 | - | 1 - 2 | 112 | 126 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK039414.1, NM_001039560.3 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
alpha-1,6-mannosyltransferase activity
alpha-1,2-mannosyltransferase activity
mannosyltransferase activity
alpha-1,3-mannosyltransferase activity
alpha-1,3-galactosyltransferase activity
molecular_function
UDP-glucose:glycoprotein glucosyltransferase activity
glycolipid mannosyltransferase activity
oligosaccharyl transferase activity
dolichyl-phosphate-glucose-glycolipid alpha-glucosyltransferase activity
acetylglucosaminyltransferase activity
acetylgalactosaminyltransferase activity
galactosyltransferase activity
fucosyltransferase activity
O antigen polymerase activity
lipopolysaccharide-1,6-galactosyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
cellulose synthase activity
9-phenanthrol UDP-glucuronosyltransferase activity
1-phenanthrol glycosyltransferase activity
9-phenanthrol glycosyltransferase activity
1,2-dihydroxy-phenanthrene glycosyltransferase activity
phenanthrol glycosyltransferase activity
beta-1,4-mannosyltransferase activity
alpha-1,2-galactosyltransferase activity
dolichyl pyrophosphate Man7GlcNAc2 alpha-1,3-glucosyltransferase activity
lipopolysaccharide-1,5-galactosyltransferase activity
dolichyl pyrophosphate Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
inositol phosphoceramide synthase activity
glucosyltransferase activity
alpha-(1->3)-fucosyltransferase activity
alpha-(1->6)-fucosyltransferase activity
indole-3-butyrate beta-glucosyltransferase activity
salicylic acid glucosyltransferase (ester-forming) activity
salicylic acid glucosyltransferase (glucoside-forming) activity
benzoic acid glucosyltransferase activity
chondroitin hydrolase activity
dolichyl-pyrophosphate Man7GlcNAc2 alpha-1,6-mannosyltransferase activity
cytokinin 9-beta-glucosyltransferase activity
biological_process
alpha-1,6-mannosyltransferase activity
alpha-1,2-mannosyltransferase activity
mannosyltransferase activity
alpha-1,3-mannosyltransferase activity
alpha-1,3-galactosyltransferase activity
molecular_function
UDP-glucose:glycoprotein glucosyltransferase activity
glycolipid mannosyltransferase activity
oligosaccharyl transferase activity
dolichyl-phosphate-glucose-glycolipid alpha-glucosyltransferase activity
acetylglucosaminyltransferase activity
acetylgalactosaminyltransferase activity
galactosyltransferase activity
fucosyltransferase activity
O antigen polymerase activity
lipopolysaccharide-1,6-galactosyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
cellulose synthase activity
9-phenanthrol UDP-glucuronosyltransferase activity
1-phenanthrol glycosyltransferase activity
9-phenanthrol glycosyltransferase activity
1,2-dihydroxy-phenanthrene glycosyltransferase activity
phenanthrol glycosyltransferase activity
beta-1,4-mannosyltransferase activity
alpha-1,2-galactosyltransferase activity
dolichyl pyrophosphate Man7GlcNAc2 alpha-1,3-glucosyltransferase activity
lipopolysaccharide-1,5-galactosyltransferase activity
dolichyl pyrophosphate Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
inositol phosphoceramide synthase activity
glucosyltransferase activity
alpha-(1->3)-fucosyltransferase activity
alpha-(1->6)-fucosyltransferase activity
indole-3-butyrate beta-glucosyltransferase activity
salicylic acid glucosyltransferase (ester-forming) activity
salicylic acid glucosyltransferase (glucoside-forming) activity
benzoic acid glucosyltransferase activity
chondroitin hydrolase activity
dolichyl-pyrophosphate Man7GlcNAc2 alpha-1,6-mannosyltransferase activity
cytokinin 9-beta-glucosyltransferase activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1962626 | NM_001039560.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1962626 | AK039414.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1962626 | AK030734.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P137990 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1962626 | NM_001039560.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1962626 | AK039414.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1962626 | AK030734.1 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864415 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864396 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864418 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864368 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864463 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864358 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864477 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864370 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864468 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864353 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864410 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864414 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864470 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864461 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864489 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864487 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864464 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864472 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864480 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864411 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864474 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21864394 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864429 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864478 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01720741 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864467 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864420 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01889067 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01473901 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02281398 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864439 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01614871 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01889062 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864500 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864501 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864493 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01473902 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01433985 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02264020 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864463 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01817928 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864449 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864455 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02281401 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864492 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864470 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864483 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864482 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864477 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01433981 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864399 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864406 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01475947 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864487 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864393 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01668198 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01473903 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864396 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01817926 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01475949 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02264012 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864450 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864427 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01481942 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864456 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02264021 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864503 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864419 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01889061 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02281403 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01668201 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01473908 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01720734 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01668199 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864431 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02210878 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01720740 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864410 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864445 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864499 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01720739 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864468 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864471 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01352320 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21864394 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10490123 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10490125 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10490124 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10490122 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10490121 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10490120 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1446314_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10490119 | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 654821_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 654821_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200045014.mm.1 | NM_001039560.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200045014.mm.1 | NM_001039560.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200045014.mm.1 | TC0200005273.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200045016.mm.1 | NM_001039560.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200045016.mm.1 | NM_001039560.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200045016.mm.1 | TC0200005273.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200045013.mm.1 | NM_001039560.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200045013.mm.1 | NM_001039560.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200045013.mm.1 | TC0200005273.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200045015.mm.1 | NM_001039560.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200045015.mm.1 | NM_001039560.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200045015.mm.1 | TC0200005273.mm.1 | MTA TC |
| Affymetrix HT Mouse Genome 430B Array | 1446314_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1446314_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3161657 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_3161657 | - | - |
| mouse-6 v1.1 (Illumina) | GI_88319967-S | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3161657 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3161657 | - | - |

Gcnt7
Genomic Map