Assay Details
| Gene Symbol: Tmem106a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144830.3 | NP_659079.1 | 1 - 2 | 86 | 61 | |
| GenBank mRNA | AK146019.1 | - | 1 - 2 | 87 | 61 | |
| AK151842.1 | - | 1 - 2 | 63 | 61 | ||
| AK154217.1 | - | 1 - 2 | 87 | 61 | ||
| AK168437.1 | - | 1 - 2 | 29 | 61 | ||
| AK169081.1 | - | 1 - 2 | 29 | 61 | ||
| AK170900.1 | - | 1 - 2 | 57 | 61 | ||
| AK172282.1 | - | 1 - 2 | 35 | 61 | ||
| BC033268.1 | - | 1 - 2 | 59 | 5037750 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK146019.1, AK151842.1, AK154217.1, AK169081.1, AK170900.1, AK172282.1, BC033268.1, NM_144830.3 transcripts.
Set Membership: |
|

Tmem106a
Genomic Map