Assay Details
| Gene Symbol: Ppp1r16b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159662.1 | NP_001153134.1 | 2 - 3 | 462 | 69 | |
| NM_153089.4 | NP_694729.1 | 2 - 3 | 440 | 69 | ||
| GenBank mRNA | AF423761.1 | - | 2 - 3 | 459 | 69 | |
| AK037627.1 | - | 757 | 69 | |||
| AK042123.1 | - | 2 - 3 | 437 | 69 | ||
| AK044453.1 | - | 2 - 3 | 478 | 69 | ||
| BC054764.1 | - | 2 - 3 | 461 | 6414610 | 69 | |
| BC057542.1 | - | 2 - 3 | 461 | 6414610 | 69 | |
| BY718617.1 | - | 2 - 3 | 462 | 69 | ||
More Information
Set Membership: |
|

Ppp1r16b
Genomic Map