Assay Details
| Gene Symbol: Arhgap40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145015.1 | NP_001138487.1 | 6 - 7 | 959 | 70 | |
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC0300002198.rn
Set Membership: |
|

Arhgap40
Genomic Map