Assay Details
| Gene Symbol: Acss2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019811.3 | NP_062785.2 | 12 - 13 | 1530 | 56 | |
| GenBank mRNA | AK028586.1 | - | 12 - 13 | 1508 | 56 | |
| AK029014.1 | - | 12 - 13 | 1498 | 56 | ||
| AK053877.1 | - | 12 - 13 | 1504 | 56 | ||
| AK093831.1 | - | 12 - 13 | 1507 | 56 | ||
| BC040790.1 | - | 12 - 13 | 1480 | 56 | ||
| BC051432.1 | - | 12 - 13 | 1530 | 6515568 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acss2
Genomic Map