Assay Details
| Gene Symbol: Slc6a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010484.2 | NP_034614.2 | 10 - 11 | 1640 | 54 | |
| GenBank mRNA | AF013604.1 | - | 9 - 10 | 1453 | 54 | |
| AK078634.1 | - | 2 - 3 | 1197 | 54 | ||
| BC108978.1 | - | 10 - 11 | 1622 | 40047534 | 54 | |
| BC108979.1 | - | 10 - 11 | 1622 | 40047537 | 54 | |
| X66119.1 | - | 9 - 10 | 1234 | 54 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc6a4
Genomic Map