Assay Details
| Gene Symbol: Slc6a4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010484.2 | NP_034614.2 | 4 - 5 | 889 | 98 | |
| GenBank mRNA | AF013604.1 | - | 3 - 4 | 702 | 98 | |
| BC108978.1 | - | 4 - 5 | 871 | 40047534 | 98 | |
| BC108979.1 | - | 4 - 5 | 871 | 40047537 | 98 | |
| X66119.1 | - | 3 - 4 | 483 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc6a4
Genomic Map