Assay Details
| Gene Symbol: Galns | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193645.1 | NP_001180574.1 | 10 - 11 | 1262 | 84 | |
| NM_016722.4 | NP_057931.3 | 12 - 13 | 1502 | 84 | ||
| GenBank mRNA | AF111346.1 | - | 12 - 13 | 1423 | 84 | |
| AK033946.1 | - | 10 - 11 | 1180 | 84 | ||
| AK159592.1 | - | 12 - 13 | 1479 | 84 | ||
| AK220245.1 | - | 12 - 13 | 1404 | 84 | ||
| BC004002.1 | - | 12 - 13 | 1356 | 3492664 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Galns
Genomic Map