Assay Details
| Gene Symbol: Galns | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193645.1 | NP_001180574.1 | 3 - 4 | 462 | 65 | |
| NM_016722.4 | NP_057931.3 | 3 - 4 | 462 | 65 | ||
| GenBank mRNA | AF111346.1 | - | 3 - 4 | 383 | 65 | |
| AK033946.1 | - | 3 - 4 | 380 | 65 | ||
| AK159592.1 | - | 3 - 4 | 439 | 65 | ||
| AK220245.1 | - | 3 - 4 | 364 | 65 | ||
| BC004002.1 | - | 3 - 4 | 316 | 3492664 | 65 | |
| BQ550288.1 | - | 3 - 4 | 228 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Galns
Genomic Map