Assay Details
| Gene Symbol: Galns | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016722.4 | NP_057931.3 | 9 - 10 | 1149 | 68 | |
| GenBank mRNA | AA793622.1 | - | 1 - 2 | 69 | 68 | |
| AF111346.1 | - | 9 - 10 | 1070 | 68 | ||
| AK159592.1 | - | 9 - 10 | 1126 | 68 | ||
| AK220245.1 | - | 9 - 10 | 1051 | 68 | ||
| BC004002.1 | - | 9 - 10 | 1003 | 3492664 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Galns
Genomic Map