Assay Details
| Gene Symbol: Cdh15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007662.2 | NP_031688.2 | 11 - 12 | 1958 | 120 | |
| GenBank mRNA | BC157978.1 | - | 11 - 12 | 1875 | 9007298 | 120 |
| M74541.1 | - | 11 - 12 | 2365 | 120 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cdh15
Genomic Map