Assay Details
| Gene Symbol: Eno3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136062.2 | NP_001129534.1 | 5 - 6 | 407 | 131 | |
| NM_001276285.1 | NP_001263214.1 | 5 - 6 | 419 | 131 | ||
| NM_007933.3 | NP_031959.1 | 5 - 6 | 519 | 131 | ||
| GenBank mRNA | AK002485.1 | - | 5 - 6 | 379 | 131 | |
| BC013460.1 | - | 4 - 5 | 391 | 4217678 | 131 | |
| CT010193.1 | - | 4 - 5 | 313 | 131 | ||
| M20745.1 | - | 2 - 3 | 139 | 131 | ||
| X57747.1 | - | 5 - 6 | 370 | 131 | ||
| X62667.1 | - | 4 - 5 | 363 | 131 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Eno3
Genomic Map