Assay Details
| Gene Symbol: Eno3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001136062.2 | NP_001129534.1 | 9 - 10 | 1161 | 81 | |
| NM_001276285.1 | NP_001263214.1 | 9 - 10 | 1173 | 81 | ||
| NM_007933.3 | NP_031959.1 | 9 - 10 | 1273 | 81 | ||
| GenBank mRNA | AK002485.1 | - | 9 - 10 | 1133 | 81 | |
| BC013460.1 | - | 8 - 9 | 1145 | 4217678 | 81 | |
| CT010193.1 | - | 8 - 9 | 1067 | 81 | ||
| M20745.1 | - | 6 - 7 | 893 | 81 | ||
| X57747.1 | - | 9 - 10 | 1124 | 81 | ||
| X62667.1 | - | 8 - 9 | 1117 | 81 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Eno3
Genomic Map