Assay Details
| Gene Symbol: Zfp276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020497.2 | NP_065243.2 | 3 - 4 | 1078 | 112 | |
| GenBank mRNA | AF178935.1 | - | 2 - 3 | 120 | 112 | |
| AK028939.1 | - | 3 - 4 | 1031 | 112 | ||
| AK043258.1 | - | 3 - 4 | 1003 | 112 | ||
| AK133363.1 | - | 3 - 4 | 1079 | 112 | ||
| BC066056.1 | - | 3 - 4 | 989 | 112 | ||
| BC078415.1 | - | 3 - 4 | 494 | 112 | ||
| BC138579.1 | - | 2 - 3 | 70 | 8861599 | 112 | |
| BC138580.1 | - | 2 - 3 | 70 | 8861600 | 112 | |
| BC145339.1 | - | 2 - 3 | 70 | 112 | ||
| BC145340.1 | - | 2 - 3 | 70 | 112 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF178935.1, BC138579.1, BC138580.1 transcripts.
Set Membership: |
|

Zfp276
Genomic Map