Assay Details
| Gene Symbol: Slc7a6os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007567.2 | NP_001007568.1 | 2 - 3 | 480 | 97 | |
| GenBank mRNA | AK134626.1 | - | 2 - 3 | 381 | 97 | |
| AK146733.1 | - | 2 - 3 | 481 | 97 | ||
| BC054118.1 | - | 2 - 3 | 478 | 30021749 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc7a6os
Genomic Map