Assay Details
| Gene Symbol: Tcp11l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146008.2 | NP_666120.1 | 8 - 9 | 1346 | 96 | |
| GenBank mRNA | AK088802.1 | - | 4 - 5 | 669 | 96 | |
| AK146035.1 | - | 8 - 9 | 1671 | 96 | ||
| BC008239.1 | - | 7 - 8 | 1142 | 96 | ||
| BC029663.1 | - | 8 - 9 | 1346 | 5364434 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tcp11l2
Genomic Map