Assay Details
| Gene Symbol: Nuak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004363.1 | NP_001004363.1 | 1 - 2 | 355 | 74 | |
| GenBank mRNA | BC082328.1 | - | 1 - 2 | 355 | 30355405 | 74 |
More Information
Updated Mapping Information
Set Membership: |
|

Nuak1
Genomic Map