Assay Details
| Gene Symbol: Nuak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004363.1 | NP_001004363.1 | 3 - 4 | 631 | 69 | |
| GenBank mRNA | AB182364.1 | - | 2 - 3 | 268 | 69 | |
| BC082328.1 | - | 3 - 4 | 631 | 30355405 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Nuak1
Genomic Map