Assay Details
| Gene Symbol: Rbl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001139516.1 | NP_001132988.1 | 3 - 4 | 573 | 64 | |
| NM_011249.2 | NP_035379.2 | 3 - 4 | 573 | 64 | ||
| GenBank mRNA | AK033402.1 | - | 3 - 4 | 567 | 64 | |
| AK147064.1 | - | 3 - 4 | 574 | 64 | ||
| AK156055.1 | - | 3 - 4 | 552 | 64 | ||
| U27177.1 | - | 3 - 4 | 557 | 64 | ||
| U27178.1 | - | 3 - 4 | 557 | 64 | ||
| U33320.1 | - | 3 - 4 | 540 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rbl1
Genomic Map