Assay Details
| Gene Symbol: Cmtm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024217.3 | NP_077179.1 | 3 - 4 | 551 | 76 | |
| GenBank mRNA | AK003882.1 | - | 3 - 4 | 406 | 77 | |
| AK079165.1 | - | 3 - 4 | 520 | 76 | ||
| AK172091.1 | - | 3 - 4 | 454 | 76 | ||
| AY241870.1 | - | 3 - 4 | 545 | 76 | ||
| BC003230.1 | - | 3 - 4 | 458 | 3483351 | 76 | |
| BC145817.1 | - | 3 - 4 | 423 | 40131120 | 76 | |
| BC145819.1 | - | 3 - 4 | 423 | 40131122 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cmtm3
Genomic Map