Assay Details
| Gene Symbol: Rrad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019662.2 | NP_062636.2 | 1 - 2 | 201 | 81 | |
| GenBank mRNA | AK077271.1 | - | 1 - 2 | 197 | 81 | |
| AK087473.1 | - | 1 - 2 | 170 | 81 | ||
| BC057138.1 | - | 1 - 2 | 180 | 5149047 | 81 | |
| BY166399.1 | - | 1 - 2 | 201 | 81 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC057138.1, NM_019662.2 transcripts.
Set Membership: |
|

Rrad
Genomic Map