Assay Details
| Gene Symbol: Cdh16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252627.1 | NP_001239556.1 | 11 - 12 | 1723 | 56 | |
| NM_001252628.1 | NP_001239557.1 | 12 - 13 | 1813 | 56 | ||
| NM_007663.3 | NP_031689.1 | 12 - 13 | 1813 | 56 | ||
| GenBank mRNA | AF016271.1 | - | 12 - 13 | 1681 | 56 | |
| AJ609635.1 | - | 12 - 13 | 1680 | 56 | ||
| AK052636.1 | - | 12 - 13 | 1616 | 56 | ||
| AK164546.1 | - | 11 - 12 | 1724 | 56 | ||
| BC015251.1 | - | 12 - 13 | 1661 | 4235757 | 56 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdh16
Genomic Map