Assay Details
| Gene Symbol: Cmtm2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028524.3 | NP_082800.1 | 1 - 2 | 275 | 73 | |
| GenBank mRNA | AK005970.1 | - | 1 - 2 | 269 | 73 | |
| AY162282.1 | - | 1 - 2 | 155 | 73 | ||
| AY241872.1 | - | 1 - 2 | 269 | 73 | ||
| BC050983.1 | - | 1 - 2 | 272 | 73 | ||
| BC060997.1 | - | 1 - 2 | 168 | 6742828 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cmtm2b
Genomic Map