Assay Details
| Gene Symbol: Tmem208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025486.2 | NP_079762.1 | 5 - 6 | 492 | 75 | |
| GenBank mRNA | AK002499.1 | - | 5 - 6 | 480 | 75 | |
| AK005662.1 | - | 5 - 6 | 476 | 75 | ||
| BC005665.1 | - | 5 - 6 | 447 | 75 | ||
| BC024608.1 | - | 5 - 6 | 443 | 4242513 | 75 | |
| BE983091.1 | - | 1 - 2 | 75 | |||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26133050 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2177944 | AK131833.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P213153 | BC005665.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P213153 | BC024608.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P213153 | BE983091.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P213153 | NM_025486.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P213153 | AK002499.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P213153 | BC005665.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P213153 | BC024608.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P213153 | BE983091.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P213153 | NM_025486.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P213153 | AK002499.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2177941 | BC005665.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2177941 | BC024608.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2177941 | NM_025486.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2177941 | AK002499.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2177941 | AK005662.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 490578 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 9044 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102662 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P101604 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P178358 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P213153 | BC005665.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P213153 | BC024608.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P213153 | BE983091.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P213153 | NM_025486.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P213153 | AK002499.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P213153 | BC005665.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P213153 | BC024608.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P213153 | BE983091.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P213153 | NM_025486.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P213153 | AK002499.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2177944 | AK131833.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2177941 | BC005665.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2177941 | BC024608.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2177941 | NM_025486.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2177941 | AK002499.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2177941 | AK005662.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26133054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26133059 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26133060 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26133052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26133070 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26133064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26133069 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26133068 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26133062 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26133051 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26133050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26133055 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26133057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26133053 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26133067 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P101604 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P102662 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P178358 | - | - |
| Whole Mouse Genome G4122A | A_51_P213153 | BC005665.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P213153 | BC024608.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P213153 | BE983091.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P213153 | NM_025486.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P213153 | AK002499.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P213153 | BC005665.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P213153 | BC024608.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P213153 | BE983091.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P213153 | NM_025486.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P213153 | AK002499.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P120213 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P101604 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P178358 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P102662 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06733009 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06732986 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06733000 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06732997 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06732989 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26133050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133062 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133054 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133053 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133051 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26133067 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06979992 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26133053 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06979999 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26133055 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06979996 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26133070 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26133051 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26133057 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26133067 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26133069 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26133064 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06733000 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26133050 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10574720 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574722 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574721 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574724 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574725 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10574719 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1419287_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10574718 | - | - |
| Mouse Genome 430 2.0 Array | 1419287_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1419287_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66320_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66320_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0800009583.mm.1 | NM_025486.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009583.mm.1 | NM_025486.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009583.mm.1 | TC0800001214.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0800009582.mm.1 | NM_025486.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009582.mm.1 | NM_025486.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0800009582.mm.1 | TC0800001214.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1419287_PM_at | - | - |
| Mouse Expression Set 430 | 1419287_at | - | - |
| Murine Genome U74v2 Set | 96342_PM_at | - | - |
| Murine Genome U74v2 Set | 96342_at | - | - |
| Murine Genome U74v2 Set | 137519_at | - | - |
| Murine Genome U74v2 Set | 137519_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419287_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1419287_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419287_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1419287_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 908939 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6110132 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006110132 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2679306 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2679306 | - | - |
| mouse-6 v1.1 (Illumina) | scl33416.4.1_0-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6110132 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl33416.4.1_0-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6110132 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2853508 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2853508 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl33416.4.1_0-S | - | - |

Tmem208
Genomic Map