Assay Details
| Gene Symbol: Cwf19l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027545.2 | NP_081821.1 | 12 - 13 | 1866 | 122 | |
| GenBank mRNA | AK033457.1 | - | 5 - 6 | 610 | 122 | |
| AK045423.1 | - | 12 - 13 | 1867 | 122 | ||
| AK083386.1 | - | 12 - 13 | 1879 | 122 | ||
| AK167727.1 | - | 5 - 6 | 589 | 122 | ||
| BC043480.1 | - | 12 - 13 | 1833 | 122 | ||
| BC125483.1 | - | 12 - 13 | 1903 | 40129998 | 122 | |
| BC125485.1 | - | 12 - 13 | 1903 | 40130000 | 122 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cwf19l2
Genomic Map