Assay Details
| Gene Symbol: Dusp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159376.1 | NP_001152848.1 | 6 - 7 | 662 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dusp15| Gene Symbol: Dusp15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159376.1 | NP_001152848.1 | 6 - 7 | 662 | 85 | |
Set Membership: |
|