Assay Details
| Gene Symbol: Mylk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081044.2 | NP_001074513.2 | 7 - 8 | 1472 | 67 | |
| GenBank mRNA | AK079396.1 | - | 7 - 8 | 1479 | 67 | |
| BC019408.1 | - | 2 - 3 | 159 | 67 | ||
| BC158041.1 | - | 7 - 8 | 1303 | 9088157 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mylk2
Genomic Map