Assay Details
| Gene Symbol: Slc35e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177766.3 | NP_808434.2 | 3 - 4 | 659 | 70 | |
| GenBank mRNA | AK031600.1 | - | 3 - 4 | 407 | 70 | |
| BC115553.1 | - | 3 - 4 | 318 | 40101801 | 70 | |
| BC115554.1 | - | 3 - 4 | 318 | 40101802 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc35e1
Genomic Map