Assay Details
| Gene Symbol: Slc35e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177766.3 | NP_808434.2 | 5 - 6 | 1023 | 63 | |
| GenBank mRNA | AK031600.1 | - | 5 - 6 | 771 | 63 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25961701 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25961702 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25961696 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25961699 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25961698 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P650325 | AK090231.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P650325 | NM_177766.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P650325 | AK090231.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P650325 | NM_177766.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2123220 | AK031600.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2123220 | NM_177766.3 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 480795 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 582859 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 560677 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 767602 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 247189 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 606813 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 674166 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 482686 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 877204 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 50585 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 156461 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P106416 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P116728 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2123220 | AK031600.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2123220 | NM_177766.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P650325 | AK090231.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P650325 | NM_177766.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P650325 | AK090231.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P650325 | NM_177766.3 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25961706 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25961699 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25961704 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25961692 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25961708 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25961695 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25961715 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25961709 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25961705 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25961698 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25961691 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25961702 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25961696 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25961703 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25961689 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25961711 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25961714 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25961687 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25961685 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P116728 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P106416 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01765745 | - | - |
| Whole Mouse Genome G4122A | A_52_P650325 | AK090231.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P650325 | NM_177766.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P650325 | AK090231.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P650325 | NM_177766.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P330677 | AK031600.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P330677 | NM_177766.3 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103162 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131322 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P116728 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P106416 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06897654 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07297610 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06897657 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06897659 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25961701 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25961702 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25961699 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25961698 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P25961696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961612 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961547 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961620 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961613 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961691 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961549 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961634 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961650 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961566 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961603 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961640 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961552 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961652 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961595 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961642 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961626 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961708 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961635 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25961555 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961711 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961692 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961695 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961629 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06878805 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07027959 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961666 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06878809 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07382285 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961662 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961679 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961650 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961696 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961689 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07297606 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961656 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961706 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961648 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961693 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07027957 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06942688 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961707 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961649 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961617 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961626 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961691 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06942646 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961620 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961651 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961668 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961631 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961655 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06878803 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07201467 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961618 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961612 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06804627 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961672 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961622 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961705 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961683 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961714 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961658 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07027963 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07247338 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07207136 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07437701 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961675 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961615 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961715 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07297610 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961613 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06896264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25961624 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10579730 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10579729 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10579725 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10579726 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10579728 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10579727 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434103_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1434104_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10579724 | - | - |
| Mouse Genome 430 2.0 Array | 1434103_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1434103_at | - | - |
| Mouse Genome 430 2.0 Array | 1434104_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1434104_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 270066_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 270066_PM_at | - | - |
| Mouse Expression Set 430 | 1434103_at | - | - |
| Mouse Expression Set 430 | 1434103_PM_at | - | - |
| Mouse Expression Set 430 | 1434104_PM_at | - | - |
| Mouse Expression Set 430 | 1434104_at | - | - |
| Murine Genome U74v2 Set | 161355_f_at | - | - |
| Murine Genome U74v2 Set | 104475_PM_at | - | - |
| Murine Genome U74v2 Set | 161355_PM_f_at | - | - |
| Murine Genome U74v2 Set | 104475_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434103_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434104_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434104_at | - | - |
| Mouse Genome 430A 2.0 Array | 1434103_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434103_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434104_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434104_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1434103_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 631065 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 4670129 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004670129 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2752618 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2752618 | - | - |
| mouse-6 v1.1 (Illumina) | scl0270066.1_219-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4670129 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl0270066.1_219-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4670129 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2752618 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2752618 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0270066.1_219-S | - | - |

Slc35e1
Genomic Map