Assay Details
| Gene Symbol: Spns3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029932.3 | NP_084208.1 | 2 - 3 | 311 | 87 | |
| GenBank mRNA | AK020683.1 | - | 2 - 3 | 312 | 87 | |
| AK156479.1 | - | 2 - 3 | 575 | 87 | ||
| AK156781.1 | - | 2 - 3 | 304 | 87 | ||
| BC150778.1 | - | 2 - 3 | 294 | 9087689 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spns3
Genomic Map