Assay Details
| Gene Symbol: Gga3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252067.1 | NP_001238996.1 | 13 - 14 | 1583 | 81 | |
| NM_173048.3 | NP_766636.2 | 14 - 15 | 1817 | 81 | ||
| GenBank mRNA | AK031086.1 | - | 14 - 15 | 1789 | 81 | |
| AK082327.1 | - | 13 - 14 | 1547 | 81 | ||
| AK122212.2 | - | 14 - 15 | 1786 | 81 | ||
| AK132183.1 | - | 14 - 15 | 1785 | 81 | ||
| AK147882.1 | - | 14 - 15 | 1785 | 81 | ||
| BC024266.1 | - | 2424 | 81 | |||
| BC025051.1 | - | 1 - 2 | 218 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gga3
Genomic Map