Assay Details
| Gene Symbol: Fdxr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007997.1 | NP_032023.1 | 8 - 9 | 900 | 67 | |
| GenBank mRNA | AK134068.1 | - | 8 - 9 | 891 | 67 | |
| BC057146.1 | - | 8 - 9 | 834 | 67 | ||
| BC071268.1 | - | 8 - 9 | 900 | 6517167 | 67 | |
| D49920.1 | - | 8 - 9 | 900 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Fdxr
Genomic Map