Assay Details
| Gene Symbol: Jmjd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033398.2 | NP_203971.2 | 4 - 5 | 1192 | 104 | |
| GenBank mRNA | AF304118.1 | - | 4 - 5 | 1125 | 104 | |
| BC006067.1 | - | 4 - 5 | 1001 | 104 | ||
| BC056629.1 | - | 4 - 5 | 1192 | 5033487 | 104 | |
| BP769727.1 | - | 2 - 3 | 104 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Jmjd6
Genomic Map