| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1100035895.mm.1 |
NM_025918.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035895.mm.1 |
NM_025918.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035895.mm.1 |
TC1100003852.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100035896.mm.1 |
NM_025918.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035896.mm.1 |
NM_025918.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035896.mm.1 |
TC1100003852.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100035894.mm.1 |
NM_025918.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035894.mm.1 |
NM_025918.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035894.mm.1 |
TC1100003852.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1100035897.mm.1 |
NM_025918.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035897.mm.1 |
NM_025918.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1100035897.mm.1 |
TC1100003852.mm.1 |
MTA TC |