Assay Details
| Gene Symbol: Ccdc43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025918.3 | NP_080194.1 | 3 - 4 | 440 | 61 | |
| GenBank mRNA | AK009319.1 | - | 3 - 4 | 436 | 61 | |
| AK011254.1 | - | 3 - 4 | 441 | 61 | ||
| AK028834.1 | - | 3 - 4 | 451 | 61 | ||
| AK161543.1 | - | 3 - 4 | 451 | 61 | ||
| AK166820.1 | - | 3 - 4 | 436 | 61 | ||
| BC058520.1 | - | 3 - 4 | 449 | 6819447 | 61 | |
| BC061076.1 | - | 3 - 4 | 449 | 6594359 | 61 | |
More Information
Set Membership: |
|

Ccdc43
Genomic Map